Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQGAP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550819

Catalog No. PA550819


Add to cart

Description

Description

The antibody detects endogenous levels of total IQGAP2 protein.

Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.
Specifications

Specifications

IQGAP2
Polyclonal
Unconjugated
IQGAP2
Liquid
IQGAP2
Synthetic peptide of human IQ motif containing GTPase activating protein 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q13576
Ras GTPase-activating-like protein IQGAP2
Rabbit
IgG
100 μL
-20°C
Primary
10788
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit