Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IQGAP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549052
Description
Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.Specifications
IQGAP2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q13576, Q3UQ44 | |
IQGAP2 | |
KLH conjugated synthetic peptide between 1436-1463 amino acids from the C-terminal region of human IQGAP2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
IQGAP2 | |
Ras GTPase-activating-like protein IQGAP2, IQGAP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10788, 544963 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction