Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK1 Recombinant Human Protein Non-distribution product as customer accommodation.

IRAK1 Recombinant Human Protein

$434.00

Specifications

Accession Number NP_001560
Assays IRAK1 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 670μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name IRAK1 Recombinant Human Protein
Concentration 0.43mg/mL
Expression System Baculovirus
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4403 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV4403
10μg Each for $434.00
Add to cart
 
Description

Description

IL-1 receptor associated kinases (IRAKs) are serine⁄threonine kinases involved in signaling via Toll⁄IL-1 pathways.
Specifications

Specifications

NP_001560
IRAK1 Recombinant Human Protein
Baculovirus
IRAK
IRAK1 Recombinant Human Protein
IRAK1 Recombinant Human Protein
95% by SDS-PAGE
440nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL
Kinase assay
GST Tag
IRAK1 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 670μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
0.43mg/mL
IRAK1, IRAK, pelle
83.4kDa
1 Each
Amino acids 194-712
For research use only. Not for use in diagnostic procedures
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit