Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK1BP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131863
Description
IRAK1BP1 Polyclonal specifically detects IRAK1BP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IRAK1BP1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| IRAK1BP1 | |
| Recombinant protein encompassing a sequence within the center region of human IRAK1BP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Monkey (97%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| ActA binding protein 3,4921528N06Rik, AIP70, interleukin-1 receptor-associated kinase 1 binding protein 1, interleukin-1 receptor-associated kinase 1-binding protein 1, MGC138458, MGC138460, SIMPL | |
| Rabbit | |
| 29 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 134728 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction