Learn More
Abnova™ IRAK3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00011213T02
Catalog No. 89-949-176
Add to cart
Specifications
Specifications
293T
NM_007199.1
pCMV-IRAK3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
IRAK3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 67.8kDa
Transient overexpression cell lysate was tested with anti-IRAK3 antibody (Mfr. No. H00011213-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human