Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK4 Rat anti-Human, Clone: 413609, R&D Systems

Rat Monoclonal Antibody

$113.85 - $319.00

Specifications

Antigen IRAK4
Clone 413609
Applications Immunohistochemistry
Host Species Rat
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB3919100 View Documents R&D Systems
MAB3919100
100 ug Each for $319.00
Add to cart
 
MAB3919SP View Documents R&D Systems
MAB3919SP
25 ug Each for $113.85
Add to cart
 
Description

Description

IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

IRAK4
Immunohistochemistry
Human
Primary
413609
Rat
Monoclonal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit