Learn More
LI-CORSupplier Diversity Partner IRDYE 680LT GOAT ANTI-HUMAN DFS Item
Manufacturer:  LI-CORSupplier Diversity Partner 92668032
IRDye 680LT Goat anti-Human IgG (H + L), 0.5 mg. Highly cross-adsorbed secondary antibody. These antibodies are not recommended when getting up and running on system. Once established near-infrared protocols are optimized with IRDye 680RD, IRDye 680LT can be used to optimize signals in the 700 channel. Dilution range 1:20,000 – 1:40,000. Note: optimization may be required with IRDye 680LT. Suitable for a variety of applications, including Western blotting, In-Gel Westerns, and many others. IMPORTANT: IRDye 680LT dye products should not be used for small animal in vivo imaging or In-Cell Western Assays.
Catalog No. NC0170408