Learn More
IREB2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 238291AP
Catalog No. 50-173-4132
Add to cart
Description
Description
Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).
Specifications
Specifications
IREB2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P48200, Q811J3
ACO3, IRE BP 2, IREB2, Iron regulatory protein 2, IRP2, IRP2AD
Rabbit
IgG
150 μL
-20°C
Primary
3658, 64602
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
IREB2
Liquid
IREB2
IREB2 Fusion Protein Ag20653
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse