Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF1 Rabbit anti-Mouse, DyLight 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178761UV

Catalog No. NBP178761UV


Add to cart

Description

Description

IRF1 Polyclonal antibody specifically detects IRF1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IRF1
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
interferon regulatory factor 1, MARIRF-1
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
3659
Western Blot
Western Blot
Affinity Purified
IRF1
The immunogen for this product maps to a region between residue 200 and 250 of mouse Interferon Regulatory Factor 1 using the numbering given in entry NP_032416.1 (GeneID 16362).
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only