Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Mouse anti-Human, Alexa Fluor 700, Clone: PCRP-IRF3-3B2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314106AF700
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
IRF3 | |
Monoclonal | |
Alexa Fluor 700 | |
interferon regulatory factor 3, IRF-3 | |
Recombinant full-length human IRF3 protein (Uniprot: Q14653) | |
0.1 mL | |
Immunology, Innate Immunity, Transcription Factors and Regulators | |
3661 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunofluorescence | |
PCRP-IRF3-3B2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title