Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF3 Mouse anti-Human, Janelia Fluor 646, Clone: 3F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104308JF646

Catalog No. NBP14308646


Add to cart

Description

Description

IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications

Specifications

IRF3
3F10
Immunocytochemistry/Immunofluorescence, Knockout Validated
Purified
IRF3
Recombinant human IRF-3(108-166aa) purified from E. coli
Protein G purified
Immunology, Innate Immunity, Transcription Factors and Regulators
Primary
3661
Human
Immunocytochemistry, Immunofluorescence, KnockDown
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
interferon regulatory factor 3, IRF-3
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-human IRF-3 mAb, clone 3F10, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF3 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit