Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3, p Ser386 Antibody (4H10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | IRF3 |
---|---|
Clone | 4H10 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, ELISASpecifications
IRF3 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
interferon regulatory factor 3, IRF-3 | |
A synthesized peptide derived from Human IRF3 [p Ser386] [UniProt Q14653] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4H10 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3661 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title