Learn More
IRF4 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF5525SP
Catalog No. AF5525SP
Add to cart
This item is not returnable. View return policy
Description
Description
IRF4 Polyclonal antibody AF5525SP specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
IRF4
LYOPH
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Q15306
interferon regulatory factor 4, IRF-4, LSIRF, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, MUM1, NF-EM5
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IRF4 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IRF3, rhIRF5, and rhIRF6 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IRF4
E. coli-derived recombinant human IRF4 Glu130-Glu451 Accession # Q15306
Immunogen affinity purified
RUO
Primary
3662
Sterile PBS to a final concentration of 0.2 mg/mL.