Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF5 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271043JF646

Catalog No. N271043J646


Add to cart

Description

Description

IRF5 Monoclonal antibody specifically detects IRF5 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

IRF5
OTI1G7
Immunohistochemistry
Affinity Purified, Conjugated
IRF5
Full length recombinant protein of human IRF5 (NP_116032) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Death Receptor Signaling Pathway
Primary
3663.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit