Learn More
IRF5 Rat anti-Mouse, Alexa Fluor 750, Clone: 903430, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB8447S100UG
Catalog No. FAB8447S100
Add to cart
This item is not returnable. View return policy
Description
Description
IRF5 Monoclonal antibody specifically detects IRF5 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
IRF5
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
P56477
IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Rat
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse IRF5 in direct ELISAs.
Flow Cytometry
903430
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Purified
IRF5
E. coli-derived recombinant mouse IRF5 Ala208-Phe288 Accession # P56477
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3663
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)