Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF6 Mouse anti-Canine, Human, Rat, Clone: OTI2C10, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503159
Description
This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.Specifications
IRF6 | |
Monoclonal | |
0.54 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O14896 | |
LPS, OFC6, PIT, PPS, PPS1, VWS, VWS1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
364081, 3664, 480015 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C10 | |
Unconjugated | |
IRF6 | |
Liquid | |
IRF6 | |
Full length human recombit protein of human IRF6 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human, Rat |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title