Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF6 Mouse anti-Human, Mouse, Rat, Canine, PerCP, Clone: OTI2C10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271044PCP
Description
IRF6 Monoclonal specifically detects IRF6 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.Specifications
IRF6 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
IRF6 | |
Full length recombinant protein of human IRF6 (NP_006138) produced in HEK293T cells. | |
0.1 mL | |
3664.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI2C10 | |
Flow Cytometry | |
interferon regulatory factor 6, IRF-6, LPS, OFC6PPS, PIT, Van der Woude syndrome, VWS, VWS1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title