Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF6 Mouse anti-Canine, Human, Rat, Clone: OTI2C10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525609

Catalog No. PIMA525609


Add to cart

Description

Description

This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
Specifications

Specifications

IRF6
Monoclonal
0.54 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O14896
Interferon regulatory factor 6, LPS, OFC6, PIT, PPS, VWS, VWS1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
364081, 3664, 480015
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2C10
Unconjugated
IRF6
Liquid
IRF6
Full length human recombit protein of IRF6 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit