Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRGM1 (LRG-47) Mouse anti-Mouse, Unconjugated, Clone: 1B2, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen IRGM1 (LRG-47)
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22641MI
View Documents
MilliporeSigma
MABF2264100UL
100 μL
Each for $372.00
Add to cart
 
MABF22642MI
View Documents
MilliporeSigma
MABF226425UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

IRGM1 (LRG-47) Monoclonal antibody specifically detects IRGM1 (LRG-47) in Mouse samples. It is validated for Immunofluorescence, Western Blotting.
Specifications

Specifications

IRGM1 (LRG-47)
Unconjugated
Mouse
Immunity-related GTPase family M protein 1;Interferon-inducible GTPase 3;Interferon-inducible protein 1;LPS-stimulated RAW 264.7 macrophage protein 47;LRG-47;LRG47
A linear peptide corresponding to 15 amino acids from the N-terminal region of mouse Immunity-related GTPase family M protein 1 (IRGM1).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunofluorescence, Western Blot
Purified
Mouse
Irgm1, Ifi1, Iigp3, Irgm
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit