Alfa Aesar™ Iridium, 1% on 5mm alumina spheres, reduced, anhydrous

Manufacturer: Alfa Aesar™ 4557009

Quantity 10g

 View more versions of this product

Catalog No. AA4557009

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011062
    Chemical Name or Material Iridium, 1% on 5mm alumina spheres, Reduced, Anhydrous

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 10g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H335
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00011062
    Chemical Name or Material Iridium, 1% on 5mm alumina spheres, Reduced, Anhydrous

    GHS H Statement
    H335
    May cause respiratory irritation.

    GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit