Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium, 99.9%, (trace metal basis), powder, ACROS Organics™

Click to view available options
Quantity:
500mg
10g
Packaging:
Glass bottle
7439-88-5
Ir
192.217
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge
23924
CHEBI:49666
iridium
[Ir]
7439-88-5
Ir
192.217
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge
23924
CHEBI:49666
iridium
[Ir]
For ultra trace analysis
Quantity 10g
Assay Percent Range 99.9%
Fieser 02,228; 09,257
Merck Index 15, 5129
Packaging Glass bottle
Formula Weight 192.2
Physical Form Amorphous Powder
Grade Trace Metal Basis
Total Trace Metal Impurities 0.1% max.
Color Gray
Chemical Name or Material Iridium
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning