Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iridium acetate, Ir 48-54%

Manufacturer:  Alfa Aesar™ H3402803

Quantity 1g

 View more versions of this product

Catalog No. AAH3402803

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Mostly used in wax oil catalytic cracking and heavy oil catalytic cracking. The catalytic water-oxidation activity of Wilkinson ft.s iridium acetate trimer has been characterized electrochemically and by using chemical oxidants. Results show it can function as an operationally homogeneous water-oxidation catalyst when driven with sodium periodate as a primary oxidant, but rapidly decomposes using Ce(IV) as a primary oxidant.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

37598-27-9
Ir(CH3COO)n
MFCD22988919
Iridium acetate

Specifications

37598-27-9
MFCD22988919
1g
Ir 48 to 54%
Ir(CH3COO)n
>100°C
Iridium acetate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H317
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P310-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit