Alfa Aesar™ Iridium acetate, Ir 48-54%

Manufacturer: Alfa Aesar™ H34028MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH34028MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 37598-27-9
    Molecular Formula Ir(CH3COO)n
    MDL Number MFCD22988919
    Chemical Name or Material Iridium acetate

    Specifications

    Molecular Formula Ir(CH3COO)n
    MDL Number MFCD22988919
    CAS 37598-27-9
    Melting Point >100°C
    Quantity 250mg
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H317
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P310-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Iridium acetate
    Assay Ir 48 to 54%

    Description

    Applications
    Mostly used in wax oil catalytic cracking and heavy oil catalytic cracking. The catalytic water-oxidation activity of Wilkinson ft.s iridium acetate trimer has been characterized electrochemically and by using chemical oxidants. Results show it can function as an operationally homogeneous water-oxidation catalyst when driven with sodium periodate as a primary oxidant, but rapidly decomposes using Ce(IV) as a primary oxidant.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H318-H317
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P310-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit