Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iridium black, 99.8% (metals basis)

Click to view available options
Quantity:
0.5g
2g
10g
7439-88-5
Ir
192.217
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge
23924
CHEBI:49666
iridium
[Ir]
7439-88-5
Ir
192.217
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge
23924
CHEBI:49666
iridium
[Ir]
UN Number UN3089
Solubility Information Soluble in aqua regia.
Physical Form S.A. >20m
Percent Purity 99.8%
Surface Area >20m2/g
Quantity 10g
Chemical Name or Material Iridium black
Assay (metals basis)

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-095-9

TSCA : Yes

Recommended Storage : Ambient temperatures