Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium carbonyl, 98+%, Acros Organics

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
18827-81-1
C12Ir4O12
18827-81-1
C12Ir4O12
Assay Percent Range 98+%
Formula Weight 1105.01
Percent Purity 98+%
Packaging Glass bottle
Solubility Solubility in water: insoluble in water.
Melting Point 230.0°C
Quantity 500mg
Chemical Name or Material Iridium carbonyl