Iridium foil, 0.25mm (0.01 in.) thick, hard, 99.8% (metals basis)

Manufacturer: Alfa Aesar 11427-FI

Quantity 50 x 50mm

 View more versions of this product

Catalog No. AA11427FI

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7439-88-5
    MDL Number MFCD00011062
    Chemical Name or Material Iridium, Temper: as rolled

    Specifications

    Quantity 50 x 50mm
    TSCA TSCA
    Physical Form Foil
    Assay Percent Range 99.8% (metals basis)
    MDL Number MFCD00011062
    CAS 7439-88-5
    Appearance White to pale yellow, foil
    Chemical Name or Material Iridium, Temper: as rolled
    Assay 99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit