Alfa Aesar™ Iridium(III) bromide hydrate, Ir 35.9% min

Manufacturer: Alfa Aesar™ 1221206

Quantity 5g

 View more versions of this product

Catalog No. AA1221206

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 431.91 (Anhydrous)
    MDL Number MFCD00049466
    Chemical Name or Material Iridium(III) bromide hydrate

    Specifications

    Formula Weight 431.91 (Anhydrous)
    Melting Point 100°C (in -3H2O)
    Physical Form Crystalline
    MDL Number MFCD00049466
    TSCA TSCA
    Quantity 5g
    Concentration Ir 35.9% min.
    Chemical Name or Material Iridium(III) bromide hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit