Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium(III) chloride trihydrate, 53-56% Ir, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13569-57-8
Cl3Ir·3H2O
352.6
LNJXVUXPFZKMNF-UHFFFAOYSA-K
iridium iii chloride trihydrate,trichloroiridium trihydrate,cl3ir.3h2o,acmc-1c5tf,iridium chloride ircl3 , trihydrate 7ci,8ci,9ci
6102153
trichloroiridium;trihydrate
O.O.O.Cl[Ir](Cl)Cl
13569-57-8
Cl3Ir·3H2O
352.6
LNJXVUXPFZKMNF-UHFFFAOYSA-K
iridium iii chloride trihydrate,trichloroiridium trihydrate,cl3ir.3h2o,acmc-1c5tf,iridium chloride ircl3 , trihydrate 7ci,8ci,9ci
6102153
trichloroiridium;trihydrate
O.O.O.Cl[Ir](Cl)Cl
Linear Formula IrCl3·3H2O
Fieser 01,132; 02,67
Merck Index 14, 5088
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in acids ans alkalies
Formula Weight 352.6
Physical Form Crystals
Percent Purity 53-56% Ir
Density 5.3000g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble in water. Other solubilities: insoluble in acids ans alkalies
Specific Gravity 5.3
Color Black to Green
Melting Point 763.0°C
Quantity 1g
Chemical Name or Material Iridium(III) chloride trihydrate
Show More Show Less