Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iridium(IV) iodide, Premion™, 99.95% (metals basis), Ir 27.0% min

Click to view available options
Quantity:
1g
5g
7790-45-6
I4Ir
699.835
MFCD00049960
BUUWLOZTIYBNKB-UHFFFAOYSA-J
iridium iv iodide,iridium iodide iri4,iridium iv iodide, premion
25113453
tetraiodoiridium
I[Ir](I)(I)I
7790-45-6
I4Ir
699.835
MFCD00049960
BUUWLOZTIYBNKB-UHFFFAOYSA-J
iridium iv iodide,iridium iodide iri4,iridium iv iodide, premion
25113453
tetraiodoiridium
I[Ir](I)(I)I
Solubility Information Insoluble in water and alcohol; Soluble in aqueous KI
Formula Weight 699.82
Physical Form Powder
Percent Purity 99.95%
Grade Premion™
Concentration Ir 27.0% min.
Quantity 1g
Chemical Name or Material Iridium(IV) iodide
Assay (metals basis)

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 232-206-3

TSCA : Yes

Recommended Storage : Ambient temperatures