Alfa Aesar™ Iridium, plasma standard solution, Specpure™, Ir 1000µg/mL

Manufacturer: Alfa Aesar™ 35751AE

Quantity 100mL

 View more versions of this product

Catalog No. AA35751AE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011062
    UN Number UN1789
    Chemical Name or Material Iridium plasma standard solution, Ir 1000μg/mL
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00011062
    Quantity 100mL
    TSCA TSCA
    UN Number UN1789
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H335
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
    Concentration IrCl3 in 20% HCl
    Chemical Name or Material Iridium plasma standard solution, Ir 1000μg/mL
    Grade Specpure™

    GHS H Statement
    H314-H335
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit