Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iridium, plasma standard solution, Specpure™, Ir 1000μg/mL

Click to view available options
Quantity:
50mL
100mL
500mL
MFCD00011062
MFCD00011062
UN Number UN1789
Concentration IrCl3 in 20% HCl
Physical Form Liquid
Grade Specpure™
Quantity 500mL
Chemical Name or Material Iridium plasma standard solution, Ir 1000μg/mL

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures