Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium standard solution, for AAS, 1 mg/ml Ir in 5% HCl, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
13569-57-8,7647-01-0,7647-01-0,7732-18-5
Cl3H6IrO3
352.612
LNJXVUXPFZKMNF-UHFFFAOYSA-K
6102153
trichloroiridium;trihydrate
O.O.O.Cl[Ir](Cl)Cl
13569-57-8,7647-01-0,7647-01-0,7732-18-5
Cl3H6IrO3
352.612
LNJXVUXPFZKMNF-UHFFFAOYSA-K
6102153
trichloroiridium;trihydrate
O.O.O.Cl[Ir](Cl)Cl
For atomic adsorption spectroscopy
Concentration 990 to 1010μg/mL of Ir(3+) in 10 to 20% HCl
Physical Form Solution
Grade AAS
Density 1.0200g/mL
Color Pink
Quantity 500mL
Chemical Name or Material Iridium standard solution

GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger