Iridium Wire, 0.25mm (0.01 in.) dia., 99.8% (Metals basis), Alfa Aesar™

Form: Approx.1.11g/m

Manufacturer: Alfa Aesar 11432BW

Quantity 50cm

 View more versions of this product

Catalog No. AA11432BW

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-88-5
    Physical Form Wire, 0.25mm (0.01 in.) dia.
    Assay Percent Range 99.8% (Metals basis)
    MDL Number MFCD00011062
    Chemical Name or Material Iridium

    Specifications

    Quantity 50cm
    Color White to pale yellow
    Physical Form Wire, 0.25mm (0.01 in.) dia.
    MDL Number MFCD00011062
    CAS 7439-88-5
    Assay Percent Range 99.8% (Metals basis)
    Appearance White to pale yellow wire
    Chemical Name or Material Iridium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit