Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Irinotecan hydrochloride trihydrate, 99%

Click to view available options
Quantity:
100mg
250mg
100286-90-6
C33H39ClN4O6
623.147
MFCD01862255
GURKHSYORGJETM-WAQYZQTGSA-N
irinotecan hydrochloride,irinotecan hcl,topotecin,campto,cpt-11,cpt 11,camptothecin 11 hydrochloride,irinotecan hydrochloride,camptothecin 11,unii-06x131e4oe
74990
CHEBI:5971
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.Cl
100286-90-6
C33H39ClN4O6
623.147
MFCD01862255
GURKHSYORGJETM-WAQYZQTGSA-N
irinotecan hydrochloride,irinotecan hcl,topotecin,campto,cpt-11,cpt 11,camptothecin 11 hydrochloride,irinotecan hydrochloride,camptothecin 11,unii-06x131e4oe
74990
CHEBI:5971
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.Cl

Topoisomerase I inhibitor

Quantity 250mg
Color Yellow
Melting Point 256°C
Merck Index 14,5091
Solubility Information Soluble in DMSO at 100mg/ml; Soluble in water at 25mg/ml with warming
Formula Weight 677.19 (623.15 Anhydrous)
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Irinotecan hydrochloride trihydrate

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DW1060750

Recommended Storage : 2° to 8°C