Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irinotecan hydrochloride trihydrate, Thermo Scientific Chemicals

Catalog Number p-7077891
Click to view available options
Quantity:
250 mg
1 g
136572-09-3
HCl·3H2O
677.18
KLEAIHJJLUAXIQ-JDRGBKBRSA-N
irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11
60837
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl
136572-09-3
HCl·3H2O
677.18
KLEAIHJJLUAXIQ-JDRGBKBRSA-N
irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11
60837
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 677.18
Percent Purity ≥96.0% (HPLC)
Chemical Name or Material Irinotecan hydrochloride trihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit