Beckman Coulter Iris™ Urine Chemistry Strips
Easy-to-use strips provide outstanding visual results
Manufacturer: HemoCue America B34886
 View more versions of this product
Catalog No. 23-043-008
  Check Availability
Add to Cart
    Specifications
    Product Type iChem 10 SG
    • Results in two minutes or less, so patients can be tested and treated in one office visit
    • Easy-to-read, flat laminated color comparison chart for increased accuracy
    • Coordinated reporting between test results pad and color chart
    • Ascorbic acid confirmation for detecting Vitamin C interference
    • Multiple test strip formats allow four different configurations for individual testing needs
    • Easy to use—simply dip, blot, read and record
    • U.S. CLIA waived
    Strip Configurations Available
    vChem 10 SG: Bilirubin, Urobilinogen, ketones, ascorbic acid, glucose, protein, blood, pH, nitrite, leukocytes, compensation, specific gravity
    vChem 8: Bilirubin, Urobilinogen, ketones, ascorbic acid, glucose, protein, blood, pH, nitrite, compensation
    vChem 6: Ketones, ascorbic acid, glucose, protein, blood, pH, leukocytes
    vChem 2: Ascorbic acid, glucose, protein
  • Description & Specifications
Related Products
Product Content Feedback