Learn More
Beckman Coulter Iris™ Urine Chemistry Strips
Easy-to-use strips provide outstanding visual results
Manufacturer:  Beckman Coulter Primary Care Diagnostics B34885
 View more versions of this product
Catalog No. 23-043-009
  Check Availability
Add to cart
Specifications
Description
  • Results in two minutes or less, so patients can be tested and treated in one office visit
  • Easy-to-read, flat laminated color comparison chart for increased accuracy
  • Coordinated reporting between test results pad and color chart
  • Ascorbic acid confirmation for detecting Vitamin C interference
  • Multiple test strip formats allow four different configurations for individual testing needs
  • Easy to use—simply dip, blot, read and record
  • U.S. CLIA waived
Strip Configurations Available
vChem 10 SG: Bilirubin, Urobilinogen, ketones, ascorbic acid, glucose, protein, blood, pH, nitrite, leukocytes, compensation, specific gravity
vChem 8: Bilirubin, Urobilinogen, ketones, ascorbic acid, glucose, protein, blood, pH, nitrite, compensation
vChem 6: Ketones, ascorbic acid, glucose, protein, blood, pH, leukocytes
vChem 2: Ascorbic acid, glucose, protein
Description & Specifications
Specifications
vChem 10 SG