Learn More
Iris Scissors
SCI_ED
Fine scissors for fine incisions.
Specifications
For Use With (Application) Surgical, Ophthalmic
Length (English) 4.3 in.
Length (Metric) 11cm
Tip Style Sharp/Sharp
View More Specs
Products 2
Catalog Number Mfr. No. Blade Style Price Quantity & Availability  
Catalog Number Mfr. No. Blade Style Price Quantity & Availability  
S17337
SCI_ED
View Documents
Medco Instruments
81450
Straight This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
S17338
SCI_ED
View Documents
Medco Instruments
81460
Curved This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
When the dissecting work calls for a light touch, reach for the Iris scissors! They have thin, stainless-steel blades with sharp points for making fine cuts.
Specifications
Specifications
Surgical, Ophthalmic
4.3 in.
11cm
Sharp/Sharp