Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Merit Iris Tissue Forceps, Sklar 4 in., 1x2 Teeth, Straight, Smooth, Thumb, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 98142
Forceps, Merit Iris Tissue Forceps, Sklar, 4 in., 1x2 Teeth, Straight, Smooth, Thumb, Stainless Steel, EA. Merit Iris Tissue Forceps are most commonly used for grasping and handling tissue in ophthalmic procedures. The narrow tips, however, allow this instrument to be beneficial for use in other microsurgical procedures as well. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with 1x2 teeth and a length of 4 inches.
Catalog No. 50-112-6703