Learn More
Alfa Aesar™ IRL-1620
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66049EXD
 View more versions of this product
Catalog No. AAJ66049EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1819.93
0.5mg
Suc-Asp-Glu-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp
-30° to -10°C
IRL-1620
HPLC
Solid
RUO
Endothelin β Receptor Agonist IRL 1620; Suc-[Glu9,Ala11,140] Endothelin-1 (8-21), human