Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(0) pentacarbonyl, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
13463-40-6
C5FeO5
Pentacarbonyl Iron
13463-40-6
C5FeO5
Pentacarbonyl Iron
Additional Information Material may have to be filtered through glass wool before use. Keep residue moist. May be pyrophoric if allowed to dry
Linear Formula Fe(CO)5
UN Number 1994
Formula Weight 195.9
Physical Form Liquid Which May Contain Particulate Matter
Density 1.4900g/mL
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: insoluble.
Specific Gravity 1.49
Boiling Point 103.0°C
Color Brown to Orange-Yellow
Melting Point -20.0°C
Quantity 1L
Flash Point −15°C
Chemical Name or Material Iron(0) Pentacarbonyl
Show More Show Less

GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Highly flammable liquid and vapour.
Fatal if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger