Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, 5% in graphite, Thermo Scientific™

Catalog Number p-7080521
Click to view available options
Quantity:
25 g
100 g
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 5% in graphite
Quantity 25 g
Physical Form Powder
UN Number UN1325
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) 5% in graphite
Chemical Name or Material Iron, Graphimet Fe-5
Hazard Category H228-H319-H335
Hazard Statement GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit