Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, 99.99%, (trace metal basis), granules, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
7439-89-6
Fe
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m
23925
CHEBI:82664
iron
[Fe]
For ultra trace analysis, Reduction reagent
Quantity 25g
Assay Percent Range 99.99%
Packaging Glass bottle
Formula Weight 55.84
Physical Form Granules
Percent Purity 99.99%
Grade Trace Metal Basis
Total Trace Metal Impurities 50ppm max.
Color Undesignated
Melting Point 1535.0°C
Boiling Point 3000.0°C
Chemical Name or Material Iron
Show More Show Less