Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, 99%, powder, -70 mesh (<212 micron), ACROS Organics™

Click to view available options
Quantity:
500g
2kg
10kg
Packaging:
Plastic bottle
7439-89-6
Fe
55.845
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.845
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m
23925
CHEBI:82664
iron
[Fe]
Reduction reagent
Quantity 2kg
Copper (Cu) 100ppm max.
Density 7.8600g/mL
Assay Percent Range 99%
Fieser 01,519; 02,229; 03,167; 05,357; 09,257; 14,184; 16,189; 17,157
Merck Index 15, 5137
Manganese (Mn) 1000ppm max.
Nickel (Ni) 500ppm max.
Packaging Plastic bottle
Formula Weight 55.84
Physical Form Fine Powder
Percent Purity 99%
Sulfur (S) 200ppm max.
Trace Analysis 50ppm max.
Color Gray
Melting Point 1535.0°C
Boiling Point 3000.0°C
Chemical Name or Material Iron
Show More Show Less