Alfa Aesar™ Iron-chrome based high temperature gas shift catalyst, HiFUEL™ W210

Manufacturer: Alfa Aesar™ 45467A4

Quantity 2.5kg

 View more versions of this product

Catalog No. AA45467A4

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Iron-chrome based high temperature gas shift catalyst HiFUEL™ W210

    Specifications

    Physical Form Pellet
    Health Hazard 2 GHS H Statement
    H334-H340-H350-H361-H373-H332-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Color Red-brown
    Quantity 2.5kg
    Particle Size 5.4mm x 3.6mm
    Health Hazard 1 Danger
    Chemical Name or Material Iron-chrome based high temperature gas shift catalyst HiFUEL™ W210

    GHS H Statement
    H334-H340-H350-H361-H373-H332-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit