Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron (Filings, Degreased), Fisher Chemical

Catalog Number i57500
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
Color Gray
Melting Point 1535°C
Boiling Point 3000°C
Atomic Weight 55.85
Density 7.86g/mL
Quantity 500 g
UN Number 3089
Identification Pass test
Formula Weight 55.85
Mesh Size 50 to 70 Mesh
Packaging Glass Bottle
Form Solid
Chemical Name or Material Iron
Show More Show Less
DOTInformation Class 4.1: Flammable Solid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit