Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Iron foil, 0.127mm (0.005 in.) thick, 99.5% (metals basis)
Click to view available options
Quantity:
25 x 25mm
25 x 100mm
Specifications
Quantity | 25 x 25mm |
Weight | ≈0.62g/25x25mm |
Physical Form | Foil |
Percent Purity | 99.5% |
Chemical Name or Material | Iron foil, 0.127mm (0.005 in.) thick |
Assay | (metals basis) |
Safety and Handling
H500
EINECSNumber : 231-096-4
TSCA : Yes
Recommended Storage : Ambient temperatures