Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iron foil, 0.25mm (0.01 in.) thick, Puratronic™, 99.995% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
7439-89-6
Fe
55.845
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.845
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum,iron, elemental,ferrous,remko,armco,ferrovac,hoeganaes eh,3zhp,ancor b,atomiron 5m
23925
CHEBI:82664
iron
[Fe]
Quantity 100 x 100mm
Weight ≈1.23g/25x25mm
Physical Form Foil
Percent Purity 99.995%
Grade Puratronic™
Chemical Name or Material Iron foil, 0.25mm (0.01 in.) thick
Assay (metals basis)