Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane

Manufacturer:  Alfa Aesar™ 40584AE

Quantity 100mL

 View more versions of this product

Catalog No. AA40584AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

331686-36-3
MFCD00798541
Iron(II) 2-ethylhexano-isopropoxide
258.14
UN1208
Iron(II) 2-ethylhexanoate monoisopropoxide

Specifications

331686-36-3
258.14
5% w/v in hexane
Iron(II) 2-ethylhexano-isopropoxide
Moisture Sensitive
Liquid
MFCD00798541
UN1208
100mL
Iron(II) 2-ethylhexanoate monoisopropoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280h-P273-P305+P351+P338-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit