Learn More
Iron(II) acetate, anhydrous, Fe 29.5% min, Thermo Scientific Chemicals
Description
Iron(II) acetate is used as a mordant by the dye industry, as a catalyst in organic oxidation reactions and in carbon nanotube synthesis. It is used as a precursor in the production of other iron compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 190°C to 200°C (decomposition) |
Color | Brown |
Physical Form | Lump/Powder |
Quantity | 25 g |
Solubility Information | Soluble in water. |
IUPAC Name | iron(2+);diacetate |
Formula Weight | 173.94 |
Odor | Characteristic |
Sensitivity | Air and moisture sensitive |
Chemical Name or Material | Iron(II) acetate, anhydrous |
Safety and Handling
Hazard Category | H315-H319-H335-H500 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 221-441-7 |
RTECSNumber | AI3850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.